Assay Details
Gene Symbol: CRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632977.3 | XP_005633034.1 | 4 - 5 | 519 | 94 | |
XM_014121923.1 | XP_013977398.1 | 4 - 5 | 526 | 94 | ||
XM_014121924.2 | XP_013977399.1 | 5 - 6 | 811 | 94 | ||
XM_022407133.1 | XP_022262841.1 | 5 - 6 | 924 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
