Assay Details
Gene Symbol: HSPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111037.2 | XP_013966512.1 | 8 - 9 | 774 | 151 | |
XM_536018.6 | XP_536018.2 | 7 - 8 | 636 | 151 | ||
XM_852308.5 | XP_857401.2 | 6 - 7 | 579 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() |
