Assay Details
Gene Symbol: RGS20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637977.3 | XP_005638034.1 | 3 - 4 | 399 | 72 | |
XM_022412315.1 | XP_022268023.1 | 4 - 5 | 1492 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
