Assay Details
Gene Symbol: ARL6IP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107937.2 | XP_013963412.1 | 836 | 82 | ||
XM_014107938.2 | XP_013963413.1 | 811 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
