Assay Details
Gene Symbol: KCNJ15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638910.3 | XP_005638967.1 | 1 - 2 | 836 | 62 | |
XM_014109822.2 | XP_013965297.1 | 2 - 3 | 259 | 62 | ||
XM_014109825.2 | XP_013965300.1 | 2 - 3 | 252 | 62 | ||
XM_014109827.2 | XP_013965302.1 | 1 - 2 | 836 | 62 | ||
XM_022413188.1 | XP_022268896.1 | 2 - 3 | 333 | 62 | ||
XM_022413189.1 | XP_022268897.1 | 2 - 3 | 282 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005638910.3, XM_014109822.2, XM_014109825.2, XM_014109827.2, XM_022413188.1, XM_022413189.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
