Assay Details
Gene Symbol: TNFAIP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626342.3 | XP_005626399.1 | 244 | 144 | ||
XM_005626344.2 | XP_005626401.1 | 472 | 144 | |||
XM_014117633.2 | XP_013973108.1 | 133 | 144 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
