Assay Details
Gene Symbol: GLRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003435454.4 | XP_003435502.1 | 5 - 6 | 946 | 125 | |
XM_022415664.1 | XP_022271372.1 | 4 - 5 | 1039 | 125 | ||
XM_852066.5 | XP_857159.2 | 5 - 6 | 1247 | 125 | ||
XM_852110.5 | XP_857203.1 | 4 - 5 | 1039 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
