Assay Details
Gene Symbol: PRKAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621218.3 | XP_005621275.1 | 967 | 105 | ||
XM_005621219.3 | XP_005621276.1 | 1029 | 105 | |||
XM_005621220.3 | XP_005621277.1 | 1006 | 105 | |||
XM_005621221.3 | XP_005621278.1 | 1016 | 105 | |||
XM_022419829.1 | XP_022275537.1 | 914 | 105 | |||
XM_022419830.1 | XP_022275538.1 | 914 | 105 | |||
XM_537920.7 | XP_537920.2 | 1012 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
