Assay Details
Gene Symbol: ELAPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621829.3 | XP_005621886.1 | 12 - 13 | 1744 | 156 | |
XM_014114773.2 | XP_013970248.1 | 12 - 13 | 1744 | 156 | ||
XM_022420361.1 | XP_022276069.1 | 12 - 13 | 1744 | 156 | ||
XM_022420362.1 | XP_022276070.1 | 12 - 13 | 1744 | 156 | ||
XM_022420363.1 | XP_022276071.1 | 12 - 13 | 1744 | 156 | ||
XM_022420364.1 | XP_022276072.1 | 12 - 13 | 1744 | 156 | ||
XM_022420365.1 | XP_022276073.1 | 12 - 13 | 1744 | 156 | ||
XM_537043.6 | XP_537043.2 | 12 - 13 | 1744 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
