Assay Details
Gene Symbol: ZNF438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617040.3 | XP_005617097.1 | 1816 | 160 | ||
XM_005617041.3 | XP_005617098.1 | 2050 | 160 | |||
XM_005617042.3 | XP_005617099.1 | 2094 | 160 | |||
XM_005617043.3 | XP_005617100.1 | 2330 | 160 | |||
XM_005617044.2 | XP_005617101.1 | 1964 | 160 | |||
XM_022406991.1 | XP_022262699.1 | 2083 | 160 | |||
XM_022406994.1 | XP_022262702.1 | 2110 | 160 | |||
XM_022406997.1 | XP_022262705.1 | 2104 | 160 | |||
XM_022406999.1 | XP_022262707.1 | 1868 | 160 | |||
XM_022407012.1 | XP_022262720.1 | 1960 | 160 | |||
XM_022407018.1 | XP_022262726.1 | 2046 | 160 |

More Information
Set Membership: |
![]() ![]() ![]() |
