Assay Details
Gene Symbol: HACD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025269.1 | NP_001020440.1 | 196 | 153 | ||
NR_131895.1 | 196 | 153 | ||||
GenBank mRNA | AJ876904.1 | - | 196 | 153 | ||
AJ876905.1 | - | 196 | 153 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
