Assay Details
Gene Symbol: TENT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627567.2 | XP_005627624.1 | 2 - 3 | 571 | 119 | |
XM_005627568.3 | XP_005627625.1 | 2 - 3 | 688 | 119 | ||
XM_014118121.2 | XP_013973596.1 | 2 - 3 | 699 | 119 | ||
XM_539017.6 | XP_539017.3 | 2 - 3 | 700 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
