Assay Details
Gene Symbol: GOLGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003432109.4 | XP_003432157.1 | 255 | 109 | ||
XM_014120091.2 | XP_013975566.1 | 223 | 109 | |||
XM_844157.5 | XP_849250.1 | 222 | 109 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
