Assay Details
Gene Symbol: CHRNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003144.2 | NP_001003144.1 | 5 - 6 | 546 | 131 | |
GenBank mRNA | AB021708.1 | - | 5 - 6 | 547 | 131 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
