Assay Details
Gene Symbol: PHYHIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618893.3 | XP_005618950.1 | 2 - 3 | 319 | 158 | |
XM_014112818.2 | XP_013968293.1 | 2 - 3 | 310 | 158 | ||
XM_022417434.1 | XP_022273142.1 | 2 - 3 | 317 | 158 | ||
XM_536357.6 | XP_536357.1 | 2 - 3 | 522 | 158 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
