Assay Details
Gene Symbol: HTR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006648.1 | NP_001006649.1 | 2 | 262 | 135 | |
XM_005640991.3 | XP_005641048.1 | 4 | 978 | 135 | ||
GenBank mRNA | AB193091.1 | - | 2 | 262 | 135 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
