Assay Details
Gene Symbol: MYCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628887.3 | XP_005628944.1 | 5 | 1039 | 117 | |
XM_005628888.3 | XP_005628945.1 | 2 | 351 | 117 | ||
XM_014119331.2 | XP_013974806.1 | 7 | 1587 | 117 | ||
XM_014119333.2 | XP_013974808.1 | 6 | 2062 | 117 | ||
XM_022427767.1 | XP_022283475.1 | 5 | 1080 | 117 | ||
XM_022427768.1 | XP_022283476.1 | 7 | 1584 | 117 | ||
XM_022427769.1 | XP_022283477.1 | 5 | 1053 | 117 | ||
XM_022427771.1 | XP_022283479.1 | 6 | 2066 | 117 | ||
XM_022427772.1 | XP_022283480.1 | 5 | 1043 | 117 | ||
XM_022427773.1 | XP_022283481.1 | 5 | 964 | 117 | ||
XM_022427774.1 | XP_022283482.1 | 4 | 646 | 117 | ||
XM_022427775.1 | XP_022283483.1 | 4 | 615 | 117 | ||
XM_022427776.1 | XP_022283484.1 | 4 | 526 | 117 | ||
XM_022427777.1 | XP_022283485.1 | 4 | 601 | 117 | ||
XM_022427778.1 | XP_022283486.1 | 3 | 532 | 117 | ||
XM_022427779.1 | XP_022283487.1 | 4 | 1510 | 117 | ||
XM_022427780.1 | XP_022283488.1 | 3 | 487 | 117 | ||
XM_022427781.1 | XP_022283489.1 | 2 | 431 | 117 | ||
XM_539578.6 | XP_539578.2 | 2 | 368 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
