Assay Details
Gene Symbol: DCAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626714.3 | XP_005626771.1 | 5 - 6 | 1080 | 111 | |
XM_022425515.1 | XP_022281223.1 | 5 - 6 | 902 | 111 | ||
XM_538706.6 | XP_538706.2 | 5 - 6 | 1393 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
