Assay Details
Gene Symbol: JMJD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639244.4 | XP_003639292.1 | 268 | 152 | ||
XM_022422697.1 | XP_022278405.1 | 267 | 152 | |||
XM_022422698.1 | XP_022278406.1 | 265 | 152 | |||
XM_022422699.1 | XP_022278407.1 | 265 | 152 | |||
XM_022422700.1 | XP_022278408.1 | 269 | 152 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
