Assay Details
Gene Symbol: MAT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003434508.4 | XP_003434556.1 | 5 - 6 | 799 | 109 | |
XM_022417726.1 | XP_022273434.1 | 5 - 6 | 799 | 109 | ||
XM_536439.5 | XP_536439.3 | 5 - 6 | 790 | 109 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
