Assay Details
Gene Symbol: ZNF567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014115501.2 | XP_013970976.2 | 7 | 1703 | 118 | |
XM_014115506.2 | XP_013970981.1 | 7 | 1971 | 118 | ||
XM_022403715.1 | XP_022259423.1 | 8 | 1805 | 118 | ||
XM_022403718.1 | XP_022259426.1 | 8 | 1835 | 118 | ||
XM_022403719.1 | XP_022259427.1 | 8 | 1801 | 118 | ||
XM_022403722.1 | XP_022259430.1 | 7 | 1640 | 118 | ||
XM_022403730.1 | XP_022259438.1 | 7 | 1728 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
