Assay Details
Gene Symbol: RHBDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419579.1 | XP_022275287.1 | 95 | 128 | ||
XM_022419580.1 | XP_022275288.1 | 95 | 128 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022419579.1, XM_022419580.1 transcripts.
Set Membership: |
![]() ![]() |
