Assay Details
Gene Symbol: TAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014119699.2 | XP_013975174.1 | 3 | 440 | 124 | |
XM_022427958.1 | XP_022283666.1 | 5 | 1259 | 124 | ||
XM_022427959.1 | XP_022283667.1 | 5 | 1306 | 124 | ||
XM_022427960.1 | XP_022283668.1 | 5 | 1252 | 124 | ||
XM_022427961.1 | XP_022283669.1 | 5 | 1194 | 124 | ||
XM_022427962.1 | XP_022283670.1 | 4 | 2317 | 124 | ||
XM_022427963.1 | XP_022283671.1 | 4 | 2264 | 124 | ||
XM_022427964.1 | XP_022283672.1 | 3 | 1558 | 124 | ||
XM_022427965.1 | XP_022283673.1 | 5 | 2790 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
