Assay Details
Gene Symbol: CPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022418861.1 | XP_022274569.1 | 4 - 5 | 334 | 135 | |
XM_022418862.1 | XP_022274570.1 | 2 - 3 | 198 | 135 | ||
XM_546705.6 | XP_546705.1 | 2 - 3 | 354 | 135 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
