Assay Details
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629868.3 | XP_005629925.1 | 3346 | 132 | ||
XM_005629869.3 | XP_005629926.1 | 3270 | 132 | |||
XM_539967.5 | XP_539967.2 | 3553 | 132 |

More Information
Set Membership: |
![]() ![]() |
