Assay Details
Gene Symbol: HMGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002937.2 | NP_001002937.1 | 320 | 185 | ||
XM_022409535.1 | XP_022265243.1 | 327 | 185 | |||
GenBank mRNA | AY135519.1 | - | 242 | 185 | ||
DN349572.1 | - | 2 | 320 | 185 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
