Assay Details
Gene Symbol: CDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639320.4 | XP_003639368.1 | 536 | 110 | ||
XM_005625479.3 | XP_005625536.1 | 536 | 110 | |||
XM_022424031.1 | XP_022279739.1 | 442 | 110 | |||
XM_022424033.1 | XP_022279741.1 | 322 | 110 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_022424033.1 transcripts.
This assay is in the 5'UTR region of the XM_022424033.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
