Assay Details
Gene Symbol: RBM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630728.3 | XP_005630785.1 | 3 - 4 | 259 | 112 | |
XM_533035.6 | XP_533035.1 | 3 - 4 | 262 | 112 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
