Assay Details
Gene Symbol: CALM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_533635.6 | XP_533635.1 | 2 - 3 | 250 | 139 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_533635.6 transcripts.
Set Membership: |
![]() ![]() ![]() |
