Assay Details
Gene Symbol: NBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624425.3 | XP_005624482.1 | 2788 | 73 | ||
XM_022422949.1 | XP_022278657.1 | 2812 | 73 | |||
XM_022422950.1 | XP_022278658.1 | 2718 | 73 | |||
XM_022422951.1 | XP_022278659.1 | 2700 | 73 | |||
XM_022422952.1 | XP_022278660.1 | 2680 | 73 | |||
XM_537628.6 | XP_537628.3 | 2694 | 73 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
