Assay Details
Gene Symbol: CDC42EP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022424306.1 | XP_022280014.1 | 1 | 1449 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CDC42EP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022424306.1 | XP_022280014.1 | 1 | 1449 | 135 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |