Assay Details
Gene Symbol: LOC100687259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639313.2 | XP_005639370.1 | 1 | 794 | 125 |
Gene Symbol: PSMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022422008.1 | XP_022277716.1 | 10 - 11 | 1258 | 125 | |
XM_535701.6 | XP_535701.2 | 12 - 13 | 1180 | 125 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
