Assay Details
Gene Symbol: DYRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625597.3 | XP_005625654.1 | 1472 | 133 | ||
XM_005625600.3 | XP_005625657.1 | 6900 | 133 | |||
XM_014117219.2 | XP_013972694.1 | 1313 | 133 | |||
XM_022424189.1 | XP_022279897.1 | 1585 | 133 | |||
XM_022424190.1 | XP_022279898.1 | 1195 | 133 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
