Assay Details
Gene Symbol: PABPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130437.2 | NP_001123909.2 | 3 - 4 | 544 | 89 | |
XM_005623168.3 | XP_005623225.1 | 3 - 4 | 520 | 89 | ||
GenBank mRNA | DN380583.1 | - | 3 - 4 | 428 | 89 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
