Assay Details
Gene Symbol: EPM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417819.1 | XP_022273527.1 | 940 | 113 | ||
XM_022417820.1 | XP_022273528.1 | 606 | 113 | |||
XM_022417821.1 | XP_022273529.1 | 937 | 113 | |||
XM_022417822.1 | XP_022273530.1 | 936 | 113 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
