Assay Details
Gene Symbol: RP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111448.2 | XP_013966923.1 | 4 - 5 | 1157 | 164 | |
XM_548972.6 | XP_548972.2 | 4 - 5 | 1158 | 164 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
