Assay Details
Gene Symbol: RXRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640843.2 | XP_005640900.1 | 5 - 6 | 992 | 185 | |
XM_014111277.1 | XP_013966752.1 | 6 - 7 | 834 | 185 | ||
XM_014111278.2 | XP_013966753.1 | 5 - 6 | 1100 | 185 | ||
XM_014111279.2 | XP_013966754.1 | 4 - 5 | 617 | 185 | ||
XM_014111280.1 | XP_013966755.1 | 4 - 5 | 614 | 185 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
