Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621941.3 | XP_005621998.1 | 8 | 4787 | 77 | |
XM_005621943.3 | XP_005622000.1 | 7 | 4644 | 77 | ||
XM_014114846.2 | XP_013970321.1 | 5 | 3876 | 77 | ||
XM_014114848.2 | XP_013970323.1 | 4 | 877 | 77 | ||
XM_022420510.1 | XP_022276218.1 | 8 | 4938 | 77 | ||
XM_022420511.1 | XP_022276219.1 | 6 | 4583 | 77 | ||
XM_022420512.1 | XP_022276220.1 | 5 | 3938 | 77 | ||
XM_022420514.1 | XP_022276222.1 | 6 | 3946 | 77 | ||
XM_022420515.1 | XP_022276223.1 | 3 | 235 | 77 | ||
XM_022420516.1 | XP_022276224.1 | 5 | 1020 | 77 | ||
XM_848131.5 | XP_853224.2 | 6 | 3999 | 77 | ||
XM_861999.5 | XP_867092.1 | 3 | 232 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
