Assay Details
Gene Symbol: MBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631969.3 | XP_005632026.1 | 15 - 16 | 5331 | 129 | |
XM_014121498.2 | XP_013976973.1 | 8 - 9 | 4545 | 129 | ||
XM_022406152.1 | XP_022261860.1 | 15 - 16 | 5331 | 129 | ||
XM_022406153.1 | XP_022261861.1 | 14 - 15 | 5206 | 129 | ||
XM_022406155.1 | XP_022261863.1 | 14 - 15 | 5277 | 129 | ||
XM_022406156.1 | XP_022261864.1 | 13 - 14 | 5232 | 129 | ||
XM_022406157.1 | XP_022261865.1 | 13 - 14 | 5089 | 129 | ||
XM_022406158.1 | XP_022261866.1 | 11 - 12 | 4990 | 129 | ||
XM_022406159.1 | XP_022261867.1 | 12 - 13 | 5035 | 129 | ||
XM_022406160.1 | XP_022261868.1 | 13 - 14 | 5103 | 129 | ||
XM_022406161.1 | XP_022261869.1 | 10 - 11 | 4868 | 129 | ||
XM_022406162.1 | XP_022261870.1 | 11 - 12 | 4861 | 129 | ||
XM_022406163.1 | XP_022261871.1 | 10 - 11 | 4816 | 129 | ||
XM_022406165.1 | XP_022261873.1 | 15 - 16 | 5331 | 129 | ||
XM_022406166.1 | XP_022261874.1 | 15 - 16 | 4632 | 129 | ||
XM_022406167.1 | XP_022261875.1 | 15 - 16 | 4632 | 129 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
