Assay Details
Gene Symbol: PGGHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631079.3 | XP_005631136.1 | 2 | 632 | 158 | |
XM_540514.5 | XP_540514.2 | 5 | 1045 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
