Assay Details
Gene Symbol: ZC3HAV1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629564.2 | XP_005629621.1 | 545 | 163 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005629564.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
