Assay Details
Gene Symbol: NIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618204.3 | XP_005618261.1 | 705 | 120 | ||
XM_005618205.3 | XP_005618262.1 | 793 | 120 | |||
XM_014112286.2 | XP_013967761.1 | 626 | 120 | |||
XM_545798.6 | XP_545798.2 | 551 | 120 |

More Information
Set Membership: |
![]() ![]() |
