Assay Details
Gene Symbol: RDH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623559.2 | XP_005623616.1 | 4 | 593 | 137 | |
XM_547866.5 | XP_547866.3 | 4 | 617 | 137 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
