Assay Details
Gene Symbol: RELL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022408770.1 | XP_022264478.1 | 953 | 125 | ||
XM_022408771.1 | XP_022264479.1 | 836 | 125 | |||
XM_022408780.1 | XP_022264488.1 | 934 | 125 | |||
XM_535220.6 | XP_535220.2 | 700 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
