Assay Details
Gene Symbol: REXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621467.3 | XP_005621524.1 | 10 - 11 | 1217 | 93 | |
XM_005621469.3 | XP_005621526.1 | 10 - 11 | 1198 | 93 | ||
XM_022420003.1 | XP_022275711.1 | 8 - 9 | 954 | 93 | ||
XM_536947.5 | XP_536947.2 | 10 - 11 | 1198 | 93 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
