Assay Details
Gene Symbol: RAB34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624865.3 | XP_005624922.1 | 2 - 3 | 435 | 117 | |
XM_005624866.3 | XP_005624923.1 | 3 - 4 | 430 | 117 | ||
XM_014116788.2 | XP_013972263.1 | 3 - 4 | 386 | 117 | ||
XM_022423348.1 | XP_022279056.1 | 2 - 3 | 160 | 117 | ||
XM_022423349.1 | XP_022279057.1 | 2 - 3 | 476 | 117 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
