Assay Details
Gene Symbol: GLIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005615924.3 | XP_005615981.1 | 3 - 4 | 751 | 87 | |
XM_005615925.3 | XP_005615982.1 | 2 - 3 | 1227 | 87 | ||
XM_005615926.3 | XP_005615983.1 | 2 - 3 | 378 | 87 | ||
XM_005615927.3 | XP_005615984.1 | 2 - 3 | 652 | 87 | ||
XM_022421731.1 | XP_022277439.1 | 6 - 7 | 2260 | 87 | ||
XM_022421732.1 | XP_022277440.1 | 7 - 8 | 2319 | 87 | ||
XM_022421733.1 | XP_022277441.1 | 7 - 8 | 2541 | 87 | ||
XM_022421734.1 | XP_022277442.1 | 6 - 7 | 1280 | 87 | ||
XM_022421735.1 | XP_022277443.1 | 7 - 8 | 2363 | 87 | ||
XM_022421739.1 | XP_022277447.1 | 8 - 9 | 2644 | 87 | ||
XM_022421740.1 | XP_022277448.1 | 7 - 8 | 1383 | 87 | ||
XM_022421745.1 | XP_022277453.1 | 5 - 6 | 2167 | 87 | ||
XM_022421750.1 | XP_022277458.1 | 5 - 6 | 938 | 87 | ||
XM_022421755.1 | XP_022277463.1 | 6 - 7 | 2269 | 87 | ||
XM_022421758.1 | XP_022277466.1 | 2 - 3 | 450 | 87 | ||
XM_022421765.1 | XP_022277473.1 | 5 - 6 | 1767 | 87 | ||
XM_022421767.1 | XP_022277475.1 | 6 - 7 | 1826 | 87 | ||
XM_541295.6 | XP_541295.3 | 3 - 4 | 1139 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
