Assay Details
Gene Symbol: ZNF598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621712.1 | XP_005621769.1 | 4 - 5 | 709 | 136 | |
XM_022420258.1 | XP_022275966.1 | 4 - 5 | 566 | 136 | ||
XM_547181.3 | XP_547181.2 | 4 - 5 | 709 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
