Assay Details
Gene Symbol: POFUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077357.1 | NP_001070825.1 | 5 - 6 | 746 | 63 | |
XM_022408821.1 | XP_022264529.1 | 6 - 7 | 847 | 63 | ||
GenBank mRNA | DQ139940.1 | - | 5 - 6 | 746 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
