Assay Details
Gene Symbol: EBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_845931.5 | XP_851024.1 | 1 - 2 | 152 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_845931.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
